Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-DAB1 (Tyr232) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104709
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104709 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104709 Supplier Invitrogen™ Supplier No. PA5104709
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of Dab1 only when phosphorylated at Tyrosine 232.

The laminar organization of multiple neuronal types in the cerebral cortex is required for normal cognitive function. In mice, the disabled-1 gene plays a central role in brain development, directing the migration of cortical neurons past previously formed neurons to reach their proper layer. This gene is similar to disabled-1, and the protein encoded by this gene is thought to be a signal transducer that interacts with protein kinase pathways to regulate neuronal positioning in the developing brain. Alternatively spliced transcript variants of this gene have been reported, but their full length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-DAB1 (Tyr232)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DAB1
Gene Accession No. O75553, P97318, Q8CJH2
Gene Alias AI956902; C630028C02Rik; Dab reelin signal transducer 1; Dab, reelin signal transducer, homolog 1; Dab, reelin signal transducer, homolog 1 (Drosophila); Dab1; DAB1, reelin adaptor protein; disabled 1; disabled homolog 1; RP23-356E21.1; scm; scr; scrambler; yot; yotari
Gene Symbols DAB1
Host Species Rabbit
Immunogen A synthesized peptide derived from human DAB1(Accession O75553), corresponding to amino acid residues around phosphorylated Tyr232.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13131, 1600, 266729
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.