Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-DAPK1 (Ser308) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564779
Description
Phospho-DAPK1 (Ser308) Polyclonal Antibody detects endogenous levels of DAPK1 only when phosphorylated at Ser308.
DAPK1 is a large, calmodulin-dependent kinase involved in gamma-interferon induced programmed cell death and is a potential tumor suppressor. DAPK1 is involved in multiple cellular signaling pathways that trigger cell survival, apoptosis, and authophagy. It regulates both type I apoptotic and type II autophagic cell death signal, depending on the cellular setting. Type I apoptotic cell death is caspase-dependent, type II authophagic cell death is caspase-independent. DAPK1 phosphorylates PIN1 resulting in inhibition of its catalytic activity, nuclear localization, and cellular function. It also phosphorylates TPM1, STX1A, PRKD1, BECN1, TSC2, RPS6, MYL9, and DAPK3.
Specifications
Phospho-DAPK1 (Ser308) | |
Polyclonal | |
Unconjugated | |
DAPK1 | |
D13Ucla1; DAP kinase 1; DAPK; DAPK1; DAP-Kinase; death associated protein kinase 1; death-associated protein kinase 1; DKFZp781I035; OTTHUMP00000216193 | |
Rabbit | |
Affinity chromatography | |
RUO | |
1612, 69635 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 30% glycerol and 0.02% sodium azide; pH 7.3 | |
P53355, Q80YE7 | |
DAPK1 | |
A synthesized peptide derived from human DAPK1 (Phospho-Ser308). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction