Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-DAPP1 (Tyr139) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121343
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121343 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121343 Supplier Invitrogen™ Supplier No. PA5121343
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The target is usually found in the following locations: Cytoplasm, Membrane, Peripheral membrane protein. Immunogen sequence: SIYES.

May act as a B-cell-associated adapter that regulates B-cell antigen receptor (BCR)-signaling downstream of PI3K.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-DAPP1 (Tyr139)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DAPP1
Gene Accession No. Q9QXT1, Q9UN19
Gene Alias b lymphocyte adapter protein Bam32; Bam32; B-cell adapter molecule of 32 kDa; Dapp1; Dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide; dual adaptor for phosphotyrosine and 3-phosphoinositides 1; dual adaptor of phosphotyrosine and 3-phosphoinositides; dual adaptor of phosphotyrosine and 3-phosphoinositides 1; hDAPP1; HSPC066; mDAPP1
Gene Symbols DAPP1
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around Y139 of human DAPP1 (NP_055210.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26377, 27071
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.