Learn More
Invitrogen™ Phospho-DARPP-32 (Thr34) Polyclonal Antibody

Description
This antibody is predicted to react with bovine, canine, chicken, human, mouse and non-human primate based on 100% sequence homology. In Western blot, this antibody detects an ~32 kDa protein representing Phospho-Thr34 DARPP-32 protein.
Specifications
Specifications
| Antigen | Phospho-DARPP-32 (Thr34) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 |
| Gene | Ppp1r1b |
| Gene Accession No. | Q60829, Q6J4I0 |
| Gene Alias | AU040756; DARPP32; DARPP-32; dopamine- and cAMP-regulated neuronal phosphoprotein; dopamine and cAMP-regulated neuronal phosphoprotein 32; dopamine- and cAMP-regulated phosphoprotein DARPP-32; FLJ20940; IPPD; neuronal phosphoprotein DARPP-32; OTTHUMP00000164275; OTTHUMP00000164276; Ppp1r1b; PPR1B; protein pho; protein phosphatase 1 regulatory inhibitor subunit 1B; protein phosphatase 1 regulatory subunit 1B; protein phosphatase 1, regulatory (inhibitor) subunit 1B |
| Gene Symbols | Ppp1r1b |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.