Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho DCC (Tyr1418) Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. ABC146625UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC1466-25UL 25 μL
ABC1466-100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABC1466-25UL Supplier MilliporeSigma Supplier No. ABC146625UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho DCC (Tyr1418) iis a rabit polyclonal antibody that detects Netrin receptor DCC and has been tested for use in Dot Blot, Immunofluorescence, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho DCC (Tyr1418)
Applications Dot Blot, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Dcc
Host Species Rabbit
Immunogen A linear peptide corresponding to 15 amino acids surrounding phospho-tyrosine 1418 from the C-terminal region of rat Netrin receptor DCC.
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_036973
Test Specificity This rabbit polyclonal antibody detects Netrin receptor DCC phosphorylalted on Tyrosine 1418 in rat cells. It targets an epitope within 15 amino acids from the C-terminal region.
Target Species Rat
Content And Storage Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.