Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-delta Catenin (Tyr904) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118687
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118687 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5118687 Supplier Invitrogen™ Supplier No. PA5118687
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of CTNND1 only when phosphorylated at Tyr904.

Delta 1 Catenin is an efficient tyrosine kinase substrate implicated both in cell transformation by SRC and in ligand-induced receptor signaling through the EGF, PDGF, CSF-1 and ERBB2 receptors. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-delta Catenin (Tyr904)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CTNND1
Gene Accession No. O60716, P30999
Gene Alias AA409437; AU019353; cadherin-associated Src substrate; Cadherin-associated Src substrate (CAS); CAS; Catenin (cadherin associated protein) delta 1; catenin (cadherin associated protein), delta 1; catenin (cadherin-associated protein), delta 1; catenin delta 1; catenin delta-1; catenin src; catenin, delta 1; Catns; CTNND; Ctnnd1; hypothetical protein; Kiaa0384; mKIAA0384; p120; p120 catenin; P120 CTN; P120 p120; P120 P120CTN; p120(CAS); p120(ctn); P120CAS; p120-catenin; P120CTN
Gene Symbols CTNND1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CTNND1(Accession O60716), corresponding to amino acid residues around phosphorylated Tyr904.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12388, 1500
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.