Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-DISC1 (Ser710) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143680
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143680 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143680 Supplier Thermo Scientific Supplier No. PA5143680
Add to Cart
Add to Cart

Phospho-DISC1 (Ser710) Polyclonal Antibody

Phospho-DISC1 (Ser710) Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-DISC1 (Ser710)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene DISC1
Gene Accession No. Q811T9
Gene Alias C1orf136; DISC1; disrupted in schizophrenia 1; disrupted in schizophrenia 1 homolog; Disrupted in schizophrenia 1 protein; KIAA0457; RP4-730B13.1; SCZD9
Gene Symbols DISC1
Host Species Rabbit
Immunogen Phospho-DISC1 (Ser-710) synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding Ser-710 in mouse DISC1. This peptide sequence is well conserved in rat DISC1 (Ser-703), but has five amino acid differences with the conserved site in human DISC1 (Ser-713).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 244667
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.