Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-DLC1 (Ser986) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567502
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567502 Supplier Invitrogen™ Supplier No. PA567502
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-DLC1 (Ser986) Polyclonal Antibody for Western Blot

This gene encodes a GTPase-activating protein that is a member of the rhoGAP family of proteins which play a role in the regulation of small GTP-binding proteins. GAP family proteins participate in signaling pathways that regulate cell processes involved in cytoskeletal changes. This gene functions as a tumor suppressor gene in a number of common cancers, including prostate, lung, colorectal, and breast cancers. Multiple transcript variants due to alternative promoters and alternative splicing have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-DLC1 (Ser986)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Dlc1
Gene Accession No. Q96QB1
Gene Alias A730069N07Rik; ARHGAP 7; ARHGAP7; deleted in liver cancer 1; deleted in liver cancer 1 protein; Deleted in liver cancer 1 protein homolog; deleted in liver cancer 1 variant 2; deleted in liver cancer variant 4; DLC 1; DLC1; Dlc-1; DLC1 Rho GTPase activating protein; DLC1 Rho GTPase activating protein; rho GTPase-activating protein 7; FLJ21120; HP; HP protein; KIAA1723; p122 RhoGAP; p122-RhoGAP; rho GTPase activating protein 7; rho GTPase-activating protein 7; RhoGAP; Rho-GTPase-activating protein 7; rho-type GTPase-activating protein 7; STARD 12; STARD12; StAR-related lipid; StAR-related lipid transfer (START) domain containing 12; stAR-related lipid transfer protein 12; START domain-containing protein 12; transfer protein 12; unm_sa1141
Gene Symbols Dlc1
Host Species Rabbit
Immunogen Synthesized peptide sequence derived from Human MLC2 around phosphorylation site of serine 986 (R-D-S(p)-G-V).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10395
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.