Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-DNA-PK (Ser2056) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121294
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121294 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121294 Supplier Invitrogen™ Supplier No. PA5121294
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa. The target is usually found in the following locations: Nucleus, nucleolus. Immunogen sequence: YSSQDP.

DNA-PK is a heterotrimeric, DNA-dependent protein kinase that initiates signaling cascades in response to DNA damage. DNA-PK is an atypical kinase, closely related to the PI3 kinases, and is often potently inhibited by small molecules designed to target PI3 kinases. DNA-PK encodes a component of the mediator complex, a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. Multiple transcript variants encoding different isoforms have been found for DNA-PK.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-DNA-PK (Ser2056)
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRKDC
Gene Accession No. P78527
Gene Alias AI326420; AU019811; DNA-dependent protein kinase catalytic subunit; DNAPDcs; DNAPK; DNA-PK catalytic subunit; DNA-PKcs; DNPK1; DOXNPH; doxorubicin nephropathy; dxnph; hyper-radiosensitivity of murine scid mutation, complementing 1; HYRC; HYRC1; I79_017613; IMD26; p350; p460; Prkdc; protein kinase, DNA activated, catalytic polypeptide; protein kinase, DNA-activated, catalytic polypeptide; scid; severe combined immunodeficiency; slip; Xrcc7
Gene Symbols PRKDC
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around S2056 of human DNA-PKcs (NP_0088355).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5591
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.