Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-DNM1L (Ser637) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101038
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101038 Supplier Invitrogen™ Supplier No. PA5101038
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of DRP1 only when phosphorylated at Ser637.

A human dynamin-related protein, DRP1 contributes to mitochondrial division in mammalian cells. It plays this important role in mitochondrial fission at steady state and during apoptosis. DRP1 is required for proper cellular distribution of mitochondria, and in mutant neurons, mitochondria are largely absent from synapses, thus providing a genetic tool to assess the role of mitochondria at synapses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-DNM1L (Ser637)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DNM1L
Gene Accession No. O00429, O35303, Q8K1M6
Gene Alias 6330417M19Rik; AI450666; C-terminal region; Dlp1; dnm1l; Dnm1p/Vps1p-like protein; Dnml1; Dnmlp1; Drp1; DVLP; DYMPLE; dynamin 1 like; dynamin 1-like; dynamin family member proline-rich carboxyl-terminal domain less; Dynamin-1-like protein; Dynamin-1-like protein-like protein; Dynamin-like protein; dynamin-like protein 1; dynamin-like protein 4; Dynamin-like protein IV; dynamin-related protein 1; DYNIV 11; DYNIV-11; EC 3.6.5.5; EGK_03514; EMPF; FLJ41912; HDYNIV; N-terminal region; python; similar to dynamin 1-like; VPS1; zgc:66163
Gene Symbols DNM1L
Host Species Rabbit
Immunogen A synthesized peptide derived from human DNM1L(Accession O00429), corresponding to amino acid residues around phosphorylated Ser637.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10059, 114114, 74006
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.