Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-DNMT1 (Ser154) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA512963
Description
Phospho-DNMT1 (Ser154) Polyclonal Antibody for DB
This gene encodes a subunit of the augmin complex, which regulates centrosome and mitotic spindle integrity, and is necessary for the completion of cytokinesis. The encoded protein was identified by interaction with ubiquitin C-terminal hydrolase 37. Alternative splicing results in multiple transcript variants.
Specifications
Phospho-DNMT1 (Ser154) | |
Polyclonal | |
Unconjugated | |
DNMT1 | |
ADCADN; AIM; CXXC9; CXXC-type zinc finger protein 9; DNA (cytosine-5-)-methyltransferase 1; DNA (cytosine-5)-methyltransferase 1; DNA methyltransferase (cytosine-5) 1; DNA methyltransferase 1; DNA methyltransferase HsaI; DNA methyltransferase I; DNA methyltransferase MmuI; DNA MTase HsaI; DNA MTase MmuI; DNA MTase RnoIP; DNMT; Dnmt1; Dnmt1o; FLJ16293; HSN1E; m.HsaI; m.MmuI; m.RnoIP; MCMT; Met1; Met-1; MGC104992; MommeD2; MTase; Uim | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
1786 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Dot Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P26358 | |
DNMT1 | |
KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S154 of human Dnmt1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction