Learn More
Invitrogen™ Phospho-DNMT1 (Ser714) Polyclonal Antibody

Description
Phospho-DNMT1 (Ser714) Polyclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | Phospho-DNMT1 (Ser714) |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | DNMT1 |
| Gene Accession No. | P26358 |
| Gene Alias | ADCADN; AIM; CXXC9; CXXC-type zinc finger protein 9; DNA (cytosine-5-)-methyltransferase 1; DNA (cytosine-5)-methyltransferase 1; DNA methyltransferase (cytosine-5) 1; DNA methyltransferase 1; DNA methyltransferase HsaI; DNA methyltransferase I; DNA methyltransferase MmuI; DNA MTase HsaI; DNA MTase MmuI; DNA MTase RnoIP; DNMT; Dnmt1; Dnmt1o; FLJ16293; HSN1E; m.HsaI; m.MmuI; m.RnoIP; MCMT; Met1; Met-1; MGC104992; MommeD2; MTase; Uim |
| Gene Symbols | DNMT1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.