Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-DOK1 (Ser450) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143682
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143682 Supplier Thermo Scientific Supplier No. PA5143682
Add to Cart
Add to Cart

Phospho-DOK1 (Ser450) Polyclonal Antibody

Phospho-DOK1 (Ser450) Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-DOK1 (Ser450)
Applications Western Blot
Classification Polyclonal
Clone WB, E
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene DOK1
Gene Accession No. P97465, Q4QQV2, Q99704
Gene Alias AW557123; Docking protein 1; docking protein 1, 62kDa (downstream of tyrosine kinase 1); Dok; DOK 1; DOK1; Dok-1; Downstream of tyrosine kinase 1; downstream of tyrosine kinase-1; MGC117395; MGC138860; p240; p62 dok; p62(dok); p62DOK; pp62; TLP1; TP1; TROVE domain family; TROVE1; VAULT2
Gene Symbols DOK1
Host Species Rabbit
Immunogen Phospho-Dok1 (Ser-450) synthetic peptide (coupled to KLH) corresponds to amino acids surrounding serine 450 in human Dok1. This sequence is conserved in Dok1 from rat (Ser-449) and mouse (Ser-451). The site is not conserved in other Dok family members.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13448, 1796, 312477
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.