Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Doublecortin (Ser47) Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS3113R

Description
In the developing cortex, cortical neurons must migrate over long distances to reach the site of their final differentiation. DCX is a cytoplasmic protein which appears to direct neuronal migration by regulating the organization and stability of microtubules. It contains two doublecortin domains, which bind microtubules. In addition, this protein interacts with LIS1, the regulatory gamma subunit of platelet activating factor acetylhydrolase, and this interaction is important to proper microtubule function in the developing cortex. Mutations in the gene encoding DCX are a cause of X-linked lissencephaly.Specifications
Phospho-Doublecortin (Ser47) | |
Polyclonal | |
Unconjugated | |
DCX | |
BOS_25702; DBCN; Dbct; DC; Dcn; Dcx; doublecortex; doublecortin; Doublin; Lissencephalin-X; lissencephaly, X-linked (doublecortin); LISX; Lis-X; LIS-XD; Neuronal migration protein doublecortin; OTTHUMP00000023860; OTTHUMP00000216316; SCLH; XLIS | |
Rabbit | |
Protein A | |
RUO | |
1641 | |
-20°C | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O43602 | |
DCX | |
KLH conjugated synthetic phosphopeptide derived from human Doublecortin around the phosphorylation site of Ser47. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction