Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-DR6 (Thr460) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105426
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105426 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105426 Supplier Invitrogen™ Supplier No. PA5105426
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of DR6 only when phosphorylated at Tyr460.

DR6 is widely expressed in human tissues and cell lines. The ligand for DR6 has not been identified. Apoptosis is induced by certain cytokines including TNF and Fas ligand of the TNF family through their death domain containing receptors, TNF-R1 and Fas. Several novel death receptors including DR3, DR4, and DR5 were recently identified. A new death domain containing receptor in the TNFR family was cloned recently and termed DR6 for death receptor-6. Like TNF-R1, DR6 interacts with death domain containing adapter molecule TRADD. Overexpression of DR6 induces apoptosis and activates NF-kappa-B and JNK.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-DR6 (Thr460)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TNFRSF21
Gene Accession No. D3ZF92, O75509
Gene Alias AA959878; BM-018; CD358; Death receptor 6; Death receptor-6; DR6; MGC31965; R74815; sCD358; sDR6; soluble CD358; soluble DR6; TNF receptor superfamily member 21; TNFR-related death receptor 6; TNFR-related death receptor-6; Tnfrsf21; TR7; Tumor necrosis factor receptor superfamily member 21; tumor necrosis factor receptor superfamily, member 21; UNQ437/PRO868
Gene Symbols TNFRSF21
Host Species Rabbit
Immunogen A synthesized peptide derived from human TNFRSF21(Accession O75509), corresponding to amino acid residues around phosphorylated Tyr460.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27242, 316256
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.