Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-DVL2 (Thr224) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988788
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8788 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8788 Supplier Bioss Supplier No. BS9060R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Disheveled DVL2 is a member of the dishevelled (dsh) protein family. The vertebrate dsh proteins have approximately 40% amino acid sequence similarity with Drosophila dsh. This gene encodes a 90-kD protein that undergoes posttranslational phosphorylation to form a 95-kD cytoplasmic protein, which may play a role in the signal transduction pathway mediated by multiple Wnt proteins. The mechanisms of dishevelled function in Wnt signaling are likely to be conserved among metazoans.

Specifications

Antigen Phospho-DVL2 (Thr224)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DVL2
Gene Accession No. O14641
Gene Alias BOS_18753; dishevelled 2; dishevelled 2 (homologous to Drosophila dsh); dishevelled 2, dsh homolog; dishevelled 2, dsh homolog (Drosophila); dishevelled segment polarity protein 2; dishevelled, dsh homolog 2; dishevelled, dsh homolog 2 (Drosophila); Dishevelled-2; DSH homolog 2; DVL2; Dvl-2; sb:eu471; Segment polarity protein dishevelled homolog DVL-2; wu:fc05d12; wu:fo71e09; wu:fp54a02; zgc:55372
Gene Symbols DVL2
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human DVL2 around the phosphorylation site of Thr224.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1856
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.