Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Dynamin 1 (Ser774) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14620
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14620 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14620 Supplier Invitrogen™ Supplier No. PA14620
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

This antibody is predicted to react with bovine, canine, chicken, human, mouse and non-human primate based on 100% sequence homology. It does not cross-react with other purified substrates of cdk5 (amphiphysin and synapsin). This antibody is specific for the ~95 kDa Dynamin protein phosphorylated at Ser774in Western blots of rat brain lysates. This antibody labels the purified protein phosphorylated in vitro by cdk5 but not by PKC. Immunolabeling is blocked by the phosphopeptide used as the antigen but not by the corresponding dephosphopeptide. Store at -20°C short term, 80°C long term.

Dynamin is a GTPase required for endocytosis of clathrin coated vesicles from the membrane. Dynamin's involvement in endocytosis was first discovered during the characterization of the temperature sensitive parylytic shibire mutation from Drosophila melanogaster. Three closely related isoforms have been identified in mammals. Dynamin1 is expressed exclusively in neurons, dynamin2 is ubiquitously expressed and dynamin3 is expressed primarily in the testes. Each of these isoforms of dynamin also has several splice variants. This suggests that each isoform/splice variant may have separate intracellular targets or function in the budding of distinct vesicle types. Common domain features of the dynamin proteins include an amino acid terminal GTPase domain, a central pleckstrin homology domain and a highly basic proline rich carboxyl terminus with several Src homology 3 (SH3) domains. These SH3 domains target the dynamin to the clathrin coated pit. Dynamin is known to interact with at least two proteins, AP2 adaptor protein complex and amphiphysin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Dynamin 1 (Ser774)
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene DNM1
Gene Accession No. P21575, P39053
Gene Alias AI838169; B-dynamin; D100; Dnm; Dnm1; Dyn1; dynamin 1; Dynamin BREDNM19; dynamin i; dynamin, brain; Dynamin1; Dynamin-1; EC 3.6.5.5; EIEE31; Ftfl; Kiaa4093; mKIAA4093; Neuronal dynamin; OTTHUMP00000215500
Gene Symbols DNM1
Host Species Sheep
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser774 of rat DNM1 conjugated to KLH.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13429, 140694
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.