Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Dynamin 1 (Ser778) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982565
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2565 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2565 Supplier Bioss Supplier No. BS13040R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Dynamin is a GTPase required for endocytosis of clathrin coated vesicles from the membrane. Dynamin′s involvement in endocytosis was first discovered during the characterization of the temperature sensitive parylytic shibire mutation from Drosophila melanogaster. Three closely related isoforms have been identified in mammals. Dynamin1 is expressed exclusively in neurons, dynamin2 is ubiquitously expressed and dynamin3 is expressed primarily in the testes. Each of these isoforms of dynamin also has several splice variants. This suggests that each isoform/splice variant may have separate intracellular targets or function in the budding of distinct vesicle types. Common domain features of the dynamin proteins include an amino acid terminal GTPase domain, a central pleckstrin homology domain and a highly basic proline rich carboxyl terminus with several Src homology 3 (SH3) domains. These SH3 domains target the dynamin to the clathrin coated pit. Dynamin is known to interact with at least two proteins, AP2 adaptor protein complex and amphiphysin.

Specifications

Antigen Phospho-Dynamin 1 (Ser778)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DNM1
Gene Accession No. Q05193
Gene Alias AI838169; B-dynamin; BOS_12077; D100; DNM; Dnm1; Dyn1; dynamin 1; Dynamin BREDNM19; Dynamin, brain; Dynamin1; Dynamin-1; EC 3.6.5.5; EIEE31; Ftfl; Kiaa4093; mKIAA4093; Neuronal dynamin; OTTHUMP00000215500
Gene Symbols DNM1
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human Dynamin 1 around the phosphorylation site of Ser778.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1759
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.