Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Dynamin (S778) Antibody, R&D Systems™

Catalog No. PPS005
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS005 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS005 Supplier R&D Systems Supplier No. PPS005
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Dynamin Polyclonal specifically detects Dynamin in Human, Mouse, Rat, Bovine, Canine, Chicken samples. It is validated for Western Blot.

Specifications

Antigen Dynamin (p ser778)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution
Gene Alias DNM, DNM1, dynamin 1, dynamin-1, EC 3.6.5.5
Gene Symbols DNM1
Host Species Sheep
Immunogen Phosphopeptide corresponding to amino acid residues surrounding the phospho-S778 of Dynamin
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1759
Test Specificity Specific for the ∼95 kDa Dynamin protein phosphorylated at S778 in Western blots. Labels the purified protein phosphorylated in vitro by Cdk5 but not by PKC. Does not cross react with other purified substrates of Cdk5 (e.g. Amphiphysin and Synapsin).
Target Species Human, Mouse, Rat, Bovine, Canine, Chicken
Content And Storage For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.