Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-DYRK1A/DYRK1B (Tyr321, Tyr273) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564574
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564574 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564574 Supplier Invitrogen™ Supplier No. PA564574
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-DYRK1A/DYRK1B (Tyr321, 273) Polyclonal Antibody detects endogenous levels of DYRK1A/DYRK1B only when phosphorylated at Tyr321, 273.

DYRK1A is a member of the dual-specificity tyrosine phosphorylation-regulated kinase (DYRK) family and is thought to be involved in neuronal differentiation, neurodegenerative diseases, and is considered to be a strong candidate gene for learning defects associated with Down syndrome. DYRK1A phosphorylates several transcription factors such as p53, leading to inhibition of embryonic neuronal cell proliferation. DYRK1A associates with multiple proteins, including SPRED1 and SPRED2; this association appears to inhibit the phosphorylation activity of DYRK1A. Recent evidence also suggests that DYRK1A may be involved in the replication of HIV-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-DYRK1A/DYRK1B (Tyr321, Tyr273)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DYRK1A
Gene Accession No. Q13627, Q61214, Q63470, Q9Y463, Q9Z188
Gene Alias 2310043O08Rik; AOMS3; D16Ertd272e; D16Ertd493e; dual specificity tyrosine phosphorylation regulated kinase 1A; dual specificity tyrosine phosphorylation regulated kinase 1B; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 1A; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 1B; dual specificity tyrosine-phosphorylation-regulated kinase 1A; Dual specificity tyrosine-phosphorylation-regulated kinase 1B; Dual Specificity Yak1-related kinase; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1A; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1b; Dyrk; DYRK1; Dyrk1a; Dyrk1b; ENSMUSG00000074897; Gm10783; hMNB; HP86; Minibrain-related kinase; MIRK; Mirk protein kinase; mmb; MNB; mnb protein kinase homolog hp86; MNB/DYRK protein kinase; MNBH; MP86; MRD7; protein kinase minibrain homolog; PSK47; RP86; serine/threonine kinase MNB; serine/threonine-specific protein kinase
Gene Symbols DYRK1A, DYRK1B
Host Species Rabbit
Immunogen A synthesized peptide derived from human DYRK1A/B (Phospho-Tyr321/273).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13548, 13549, 1859, 25255, 308468, 9149
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.