Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-E2F1 (Ser375) Rabbit anti-Human, Unconjugated, Clone: 71-10, MilliporeSigma™

Catalog No. MABE178225 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1782-25 25 μL
MABE1782-100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE1782-25 Supplier MilliporeSigma Supplier No. MABE178225UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Anti-phospho-E2F1 (Ser375), clone 71-10, Cat. No. MABE1782, is a highly specific rabbit monoclonal antibody that targets Transcription factor E2F1 phosphorylated on Serine 375 and has been tested in ELISA and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-E2F1 (Ser375)
Applications Western Blot, ELISA
Classification Monoclonal
Clone 71-10
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Transcription factor E2F1;E2F-1;PBR3;Retinoblastoma-associated protein 1;RBAP-1;Retinoblastoma-binding protein 3;RBBP-3;pRB-binding protein E2F-1
Gene Symbols E2F1;RBBP3
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 17 amino acids surrounding phosphoserine 375 from the C-terminal region of human transcription factor E2F1.
Molecular Weight of Antigen ∽80 kDa observed ( GST fusion protein); 46.92 kDa calculated (without GST). Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_005216.1
Test Specificity Clone 71-10 is a rabbit monoclonal antibody that specifically detects human E2F1 phosphorylated on Serine 375.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.