Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-E4B (Ser504) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986503
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6503 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6503 Supplier Bioss Supplier No. BS5346R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The eukaryotic translation initiation factor 4B (eIF4B) plays a critical role in recruiting the 40S ribosomal subunit to the mRNA. It functions in close association with eIF4F and eIF4A. It binds near the 5′-terminal cap of mRNA in the presence of eIF4F and ATP. It promotes the ATPase activity and the ATP-dependent RNA unwinding activity of both eIF4A and eIF4F.

Specifications

Antigen Phospho-E4B (Ser504)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Eif4b
Gene Accession No. P23588
Gene Alias 2310046H11Rik; AL024095; BOS_5043; C85189; Eif4a2; EIF4B; eIF-4B; Eukaryotic translation initiation factor 4B; hCG_2014609; LOC100354421; PRO1843
Gene Symbols Eif4b
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human EIF4B around the phosphorylation site of Ser504.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1975
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.