Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-ECT2 (Thr790)Rabbit anti-Canine, Chicken, Chimpanzee, Human, Mouse, Rat, Zebrafish, Polyclonal, Rockland™
SDP

Catalog No. 501991877
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1877 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1877 Supplier Rockland Immunochemicals Supplier No. 600401935
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity-purified antibody is directed against the phosphorylated form of human ECT2 protein at the pT790 residue. Reactivity occurs against human ECT2 protein at the pT790 residue. By ELISA the antibody is specific for the phosphorylated form of the protein. Reactivity with non-phosphorylated human ECT2 is minimal by ELISA, but western blot shows specific staining of interphase cell lysates, which are predicted to contain non-phosphorylated ECT2. Additional experiments are underway to clarify antibody specificity. A BLAST analysis was used to suggest cross reactivity with ECT2 protein from human, mouse, rat, zebrafish, chimpanzee, chicken, and dog based on 100% homology with the immunizing sequence. Reactivity against homologues from other sources is not known.

The protein encoded by this gene is a transforming protein that is related to Rho-specific exchange factors and yeast cell cycle regulators. The expression of this gene is elevated with the onset of DNA synthesis and remains elevated during G2 and M phases. In situ hybridization analysis showed that expression is at a high level in cells undergoing mitosis in regenerating liver. Thus, this protein is expressed in a cell cycle-dependent manner during liver regeneration, and is thought to have an important role in the regulation of cytokinesis.

Specifications

Antigen Phospho-ECT2 (Thr790)
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene ECT2
Gene Accession No. Q07139, Q9H8V3
Gene Alias AI528536; ARHGEF31; BOS_1061; Ect2; ect2 oncogene; ect2 protein; epithelial cell transforming 2; epithelial cell transforming sequence 2 oncogene; epithelial cell transforming sequence 2 oncogene protein; Epithelial cell-transforming sequence 2 oncogene; FLJ10461; LOW QUALITY PROTEIN: protein ECT2; MGC138291; mKIAA4037; Protein ECT2
Gene Symbols ECT2
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13605, 1894, 361921, 424983, 445406, 460853, 488172
Target Species Canine, Chicken, Chimpanzee, Human, Mouse, Rat, Zebrafish
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.