Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-EEF1A2 (Ser358) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535404
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535404 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535404 Supplier Invitrogen™ Supplier No. PA535404
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with chicken, non-human primate, rat, sheep and Xenopus based on 100% sequence homology. This antibody contains enough material to conduct 10 mini-Western blots.

This gene encodes an isoform of the alpha subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This isoform (alpha 2) is expressed in brain, heart and skeletal muscle, and the other isoform (alpha 1) is expressed in brain, placenta, lung, liver, kidney, and pancreas. This gene may be critical in the development of ovarian cancer. [provided by RefSeq, Mar 2014].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-EEF1A2 (Ser358)
Applications Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 0.1mg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene EEF1A2
Gene Accession No. P62631, Q05639
Gene Alias Eef1a; EEF1A2; eEF1A-2; EEF1AL; EF1A; EF-1-alpha-2; EIEE33; elongation factor 1 A-2; elongation factor 1-alpha 2; Eukaryotic elongation factor 1 A-2; eukaryotic translation elongation factor 1 alpha 2; HS1; MRD38; Ps10; RATPS10; S1; statin S1; Statin-like protein; statin-S1; STN; Stnl; wasted; wst
Gene Symbols EEF1A2
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser358 of human eEF1A2 conjugated to KLH.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13628, 1917
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.