Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-EEF2 (Tyr443) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105490
Description
Antibody detects endogenous levels of EEF2 only when phosphorylated at Tyr443.
Catalyzes the GTP-dependent ribosomal translocation step during translation elongation. During this step, the ribosome changes from the pre-translocational (PRE) to the post-translocational (POST) state as the newly formed A-site-bound peptidyl-tRNA and P-site-bound deacylated tRNA move to the P and E sites, respectively. Catalyzes the coordinated movement of the two tRNA molecules, the mRNA and conformational changes in the ribosome.
Specifications
| Phospho-EEF2 (Tyr443) | |
| Polyclonal | |
| Unconjugated | |
| EEF2 | |
| EEF2; EEF-2; Ef2; EF-2; Elongation factor 2; elongation factor-2; eukaryotic translation elongation factor 2; I79_013848; polypeptidyl-tRNA translocase; SCA26; zef2 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 13629, 1938, 29565 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P05197, P13639, P58252 | |
| EEF2 | |
| A synthesized peptide derived from human EEF2(Accession P13639), corresponding to amino acid residues around phosphorylated Tyr443. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction