Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-EGFR (Tyr1173) Mouse anti-Human, Clone: 3E7.2, MilliporeSigma™

Catalog No. MABS829MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS829MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS829MI Supplier MilliporeSigma Supplier No. MABS829
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect phospho EGFR using this mouse monoclonal Anti-phospho-EGFR (Tyr1173), clone 3E7.2 used in Immunocytochemistry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-EGFR (Tyr1173)
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 3E7.2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols EGFR,ERBB,ERBB1,HER1
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 13 amino acids surrrounding Tyrosine 1173 of human epidermal growth factor receptor.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_005219
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.