Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-eIF2 beta (Tyr298) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105524
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105524 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105524 Supplier Thermo Scientific Supplier No. PA5105524
Add to Cart
Add to Cart

Phospho-eIF2 beta (Tyr298) Polyclonal Antibody

Phospho-eIF2 beta (Tyr298) Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-eIF2 beta (Tyr298)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EIF2S2
Gene Accession No. P20042, Q99L45
Gene Alias 2810026E11Rik; 38kDa; AA408636; AA571381; AA986487; AW822225; D2Ertd303e; EIF2; EIF2B; EIF2BA; EIF2beta; eIF-2-beta; EIF2S2; Eukaryotic translation initiation factor 2 subunit 2; eukaryotic translation initiation factor 2 subunit beta; eukaryotic translation initiation factor 2, subunit 2 (beta); eukaryotic translation initiation factor 2, subunit 2 (beta, 38kDa); eukaryotic translation initiation factor 2, subunit 2 beta; eukaryotic translation initiation factor 2, subunit 2 beta, 38kDa; PPP1R67; protein phosphatase 1, regulatory subunit 67
Gene Symbols EIF2S2
Host Species Rabbit
Immunogen A synthesized peptide derived from human EIF2S2(Accession P20042), corresponding to amino acid residues around phosphorylated Tyr298.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296302, 67204, 8894
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.