Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-EIF2S1 (Ser51) Monoclonal Antibody (1A4A11), Proteintech
SDP

Catalog No. 502333515 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-3515 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-3515 Supplier Proteintech Group Inc Supplier No. 680231IG100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Phospho-EIF2S1 (Ser51) Monoclonal antibody specifically detects Phospho-EIF2S1 (Ser51) in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Phospho-EIF2S1 (Ser51)
Applications Western Blot
Classification Monoclonal
Clone 1A4A11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Accession No. P05198
Gene Alias 0910001O23Rik; 2410026C18Rik; 35kDa; EIF-2; EIF-2A; EIF-2alpha; EIF2; EIF2A; eIF2alpha; EIF2S1
Host Species Mouse
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1965
Target Species Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.