Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-eIF4E (Ser209) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585816
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585816 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585816 Supplier Invitrogen™ Supplier No. PA585816
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Rabbit (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Positive Control: Treated HCT116, Treated HeLa, C2C12. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

eIF4E is involved in several cellular processes including enhanced translational efficiency, splicing, mRNA stability, and RNA nuclear export. All eukaryotic cellular mRNAs are blocked at their 5-prime ends with the 7-methylguanosine cap structure, m7GpppX (where X is any nucleotide). EIF4E is a eukaryotic translation initiation factor involved in directing ribosomes to the cap structure of mRNAs. It is a 24-kD polypeptide that exists as both a free form and as part of a multiprotein complex termed EIF4F. The EIF4E polypeptide is the rate-limiting component of the eukaryotic translation apparatus and is involved in the mRNA-ribosome binding step of eukaryotic protein synthesis. The other subunits of EIF4F are a 50-kD polypeptide, termed EIF4A, that possesses ATPase and RNA helicase activities, and a 220-kD polypeptide, EIF4G.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-eIF4E (Ser209)
Applications Western Blot
Classification Polyclonal
Concentration 1.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene Eif4e
Gene Accession No. P06730, P63073
Gene Alias AUTS19; CBP; EG668879; EIF4E; EIF-4E; EIF4E1; EIF4EL1; Eif4e-ps; EIF4F; eIF-4F 25 kDa subunit; eukaryotic translation initiation factor 4E; eukaryotic translation initiation factor 4E-like 1; eukaryotic translation initiation factor 4E-like protein; eukaryotic translation initiation factor 4-like protein; If4e; MGC111573; mRNA cap-binding protein; OTTHUMP00000219703; OTTHUMP00000219704; OTTHUMP00000219705; translation initiation factor eIF-4E
Gene Symbols Eif4e
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide surrounding phospho Ser209 of human eIF4E. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13684, 1977
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.