Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-eIF4E (Ser209) Recombinant Rabbit Monoclonal Antibody (RM452)

Rabbit Recombinant Monoclonal Antibody

Supplier:  Invitrogen™ MA562157

Encompass_Preferred

Catalog No. MA562157


Only null left
Add to Cart

Description

Description

This antibody reacts to human eIF4E only when phosphorylated at Ser209. There is no cross-reactivity to eIF4E that is not phosphorylated. This antibody may also react to mouse or rat Phospho-eIF4E (Ser209) as predicted by immunogen homology.

eIF4E is involved in several cellular processes including enhanced translational efficiency, splicing, mRNA stability, and RNA nuclear export. All eukaryotic cellular mRNAs are blocked at their 5-prime ends with the 7-methylguanosine cap structure, m7GpppX (where X is any nucleotide). EIF4E is a eukaryotic translation initiation factor involved in directing ribosomes to the cap structure of mRNAs. It is a 24-kD polypeptide that exists as both a free form and as part of a multiprotein complex termed EIF4F. The EIF4E polypeptide is the rate-limiting component of the eukaryotic translation apparatus and is involved in the mRNA-ribosome binding step of eukaryotic protein synthesis. The other subunits of EIF4F are a 50-kD polypeptide, termed EIF4A, that possesses ATPase and RNA helicase activities, and a 220-kD polypeptide, EIF4G.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-eIF4E (Ser209)
Recombinant Monoclonal
Conc. Not Determined
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
P06730
Eif4e
A phospho-peptide corresponding to human Phospho-eIF4E (Ser209).
100 μL
Primary
Human
Antibody
IgG
Western Blot
RM452
Unconjugated
Eif4e
AUTS19; CBP; EG668879; EIF4E; EIF-4E; EIF4E1; EIF4EL1; Eif4e-ps; EIF4F; eIF-4F 25 kDa subunit; eukaryotic translation initiation factor 4E; eukaryotic translation initiation factor 4E-like 1; eukaryotic translation initiation factor 4E-like protein; eukaryotic translation initiation factor 4-like protein; If4e; MGC111573; mRNA cap-binding protein; OTTHUMP00000219703; OTTHUMP00000219704; OTTHUMP00000219705; translation initiation factor eIF-4E
Rabbit
Protein A
RUO
1977
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.