Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-EIF6 (Ser235) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106173
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106173 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106173 Supplier Invitrogen™ Supplier No. PA5106173
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of EIF6 only when phosphorylated at Ser235.

Hemidesmosomes are structures which link the basal lamina to the intermediate filament cytoskeleton. An important functional component of hemidesmosomes is the integrin beta-4 subunit (ITGB4), a protein containing two fibronectin type III domains. The protein encoded by this gene binds to the fibronectin type III domains of ITGB4 and may help link ITGB4 to the intermediate filament cytoskeleton. The encoded protein, which is insoluble and found both in the nucleus and in the cytoplasm, can function as a translation initiation factor and prevent the association of the 40S and 60S ribosomal subunits. Multiple transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-EIF6 (Ser235)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Eif6
Gene Accession No. O55135, P56537, Q3KRD8
Gene Alias 1110004P11; AA408895; b(2)gcn; B(2)GCN homolog; B4 integrin interactor; CAB; EIF3A; Eif6; eIF-6; eIF-6 {ECO:0000255; eukaryotic translation initiation factor 3A; eukaryotic translation initiation factor 6; eukaryotic translation initiation factor 6 {ECO:0000255; HAMAP-Rule:MF_03132}; imc-415; integrin beta 4 binding protein; Itgb4bp; OK/SW-cl.27; p27 beta-4 integrin-binding protein; p27(BBP); p27BBP
Gene Symbols Eif6
Host Species Rabbit
Immunogen A synthesized peptide derived from human EIF6(Accession P56537), corresponding to amino acid residues around phosphorylated Ser235.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16418, 305506, 3692
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.