Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho Enol Pyruvate CarboxylaseRabbit anti-Plant, Polyclonal, Rockland™
SDP

Catalog No. 501990178
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0178 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-0178 Supplier Rockland Immunochemicals Supplier No. 1004163
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-rabbit serum, purified and partially purified Phospho-enol-pyruvate Carboxylase [Maize Leaves]. Cross reactivity against Phospho-enol-pyruvate Carboxylase from other tissues and species may occur but have not been specifically determined.

Phosphoenolpyruvate Carboxylase, through the carboxylation of phosphoenolpyruvate (PEP), forms an oxaloacetate, a four-carbon dicarboxylic acid source for the tricarboxylic acid cycle. Phospho-enol-pyruvate Carboxylase enzyme is regulated by light-reversible phosphorylation. This protein is involved in the pathway C3 acid pathway, which is part of Photosynthesis.

Specifications

Antigen Phospho Enol Pyruvate Carboxylase
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 90 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Host Species Rabbit
Immunogen Phospho-enol-pyruvate Carboxylase [Maize Leaves]
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.