Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-eNOS/nNOS (Tyr657, Tyr895) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143729
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143729 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143729 Supplier Invitrogen™ Supplier No. PA5143729
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects a 140 kDa* band on SDS-PAGE immunoblots of human umbilical vein endothelial cells treated with pervanadate, and this reactivity is not observed after akaline phosphatase treatment.

Nitric oxide (NO) has a broad range of biological activities and is implicated in signaling pathways in phylogenetically diverse species. Nitric oxide synthases (NOS), the enzymes responsible for synthesis of NO, are homodimers whose monomers are themselves two fused enzymes: a cytochrome reductase and a cytochrome that requires three cosubstrates (L-arginine, NADPH, and oxygen) and five cofactors or prosthetic groups (FAD, FMN, calmodulin, tetrahydrobiopterin, and heme). Several distinct NOS isoforms are produced from three distinct genes. The inducible form of NOS, iNOS (NOS-II), is Ca2+ independent and is expressed in a broad range of cell types, and two constitutive Ca2+/CaM-dependent forms of NOS: nNOS (bNOS, NOS-I) identified in neurons and eNOS (ecNOS, NOS-III) identified in endothelial cells. Regulation of eNOS activity occurs through phosphorylation at multiple sites. Phosphorylation of Ser-633 (mouse Ser-632) in the FMN binding domain increases eNOS activity and may be important for the maintenance of NO synthesis after initial activation by Ca2+ flux and Ser-1177 phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-eNOS/nNOS (Tyr657, Tyr895)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration ∼1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene NOS3
Gene Accession No. P29474, P70313, Q62600
Gene Alias endothelial Nitric Oxide Synthase; eNOS; NOS-III
Gene Symbols NOS3
Host Species Rabbit
Immunogen Phospho-eNOS (Tyr-657) synthetic peptide (coupled to carrier protein) corresponding to amino acids surrounding tyrosine 657 in human eNOS. This sequence is conserved in mouse (Tyr-656) and rat (Tyr-656) eNOS, and is identical to the conserved site in nNOS (Tyr-895). The site is also well conserved in iNOS (Tyr-631).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18127, 24600, 4846
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.