Learn More
Invitrogen™ Phospho-eNOS (Ser1179) Polyclonal Antibody

Description
Antibody detects endogenous levels of eNOS only when phosphorylated at Ser1179.
Specifications
Specifications
| Antigen | Phospho-eNOS (Ser1179) |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | NOS3 |
| Gene Accession No. | P29474, P70313, Q62600 |
| Gene Alias | 2310065A03Rik; cNOS; Constitutive NOS; Ecnos; EC-NOS; endothelial nitric oxide synthase; endothelial nitric oxide synthase 3; endothelial nitric oxide synthase NOS3; endothelial NOS; eNos; e-NOS; nitric oxide synthase; nitric oxide synthase 3; nitric oxide synthase 3 (endothelial cell); nitric oxide synthase 3 transcript variant eNOS-delta20; nitric oxide synthase 3 transcript variant eNOS-delta20-21; nitric oxide synthase 3 transcript variant eNOS-delta21; nitric oxide synthase 3, endothelial cell; nitric oxide synthase, endothelial; NOS; NOS type III; Nos3; Nos-3; NOSIII; OTTHUMP00000213183 |
| Gene Symbols | NOS3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.