Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-eNOS (Ser602) Rabbit anti-Bovine, Polyclonal, MilliporeSigma™

Catalog No. ABS1631MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1631MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1631MI Supplier MilliporeSigma Supplier No. ABS1631
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect endothelial Nitric oxide synthase using this rabbit polyclonal Anti-phospho-eNOS (Ser602). It has been tested for use in ELISA and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-eNOS (Ser602)
Applications Western Blot, ELISA
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols NOS3
Host Species Rabbit
Immunogen A linear peptide of 11 amino acids from the N-terminal edge of the 42-residue bovine eNOS autoinhibitory element.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_851380
Test Specificity This polyclonal antibody specifically detects the phospho-Ser602 in eNOS. It targets a sequence of 11 amino acids surrounding Ser602.
Target Species Bovine
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.