Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-eNOS (Ser633) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564551
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564551 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564551 Supplier Invitrogen™ Supplier No. PA564551
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-eNOS (Ser633) Polyclonal Antibody detects endogenous levels of eNOS only when phosphorylated at Ser633.

Nitric oxide (NO) is an inorganic, gaseous free radical that carries a variety of messages between cells. Vasorelaxation, neurotransmission and cytotoxicity can all be potentiated through cellular response to NO. NO production is mediated by members of the nitric oxide synthase (NOS) family. NOS catalyzes the oxidization of L-arginine to produce L-citrulline and NO. Two constitutive isoforms, brain or neuronal NOS (b or nNOS, type I) and endothelial cell NOS (eNOS, type III), and one inducible isoform (iNOS, type II), have been cloned. All NOS isoforms contain calmodulin, nicotinamide adenine dinucleotide phosphate (NADPH), flavin adenine dinucleotide (FAD), and flavin mononucleotide (FMN) binding domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-eNOS (Ser633)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NOS3
Gene Accession No. P29474, P70313, Q62600
Gene Alias 2310065A03Rik; cNOS; Constitutive NOS; Ecnos; EC-NOS; endothelial nitric oxide synthase; endothelial nitric oxide synthase 3; endothelial nitric oxide synthase NOS3; endothelial NOS; eNos; e-NOS; nitric oxide synthase; nitric oxide synthase 3; nitric oxide synthase 3 (endothelial cell); nitric oxide synthase 3 transcript variant eNOS-delta20; nitric oxide synthase 3 transcript variant eNOS-delta20-21; nitric oxide synthase 3 transcript variant eNOS-delta21; nitric oxide synthase 3, endothelial cell; nitric oxide synthase, endothelial; NOS; NOS type III; Nos3; Nos-3; NOSIII; OTTHUMP00000213183
Gene Symbols NOS3
Host Species Rabbit
Immunogen A synthesized peptide derived from human eNOS (Phospho-Ser633).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18127, 24600, 4846
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.