Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-EPH A3/A4/A5 (Tyr779) Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501987828
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7828 Supplier Bioss Supplier No. BS7555R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EPHA3 (EPA receptor A3), also known as ETK, HEK, ETK1, HEK4, TYRO4, is belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EPHA3 binds ephrin-A ligands.

Specifications

Antigen Phospho-EPH A3/A4/A5 (Tyr779)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EPHA3
Gene Accession No. O08680, P29319, P29320
Gene Alias AW492086; Cek4; EK4; End3; Eph receptor A3; Epha3; EPH-like kinase 4; Eph-like tyrosine kinase 1; Ephrin type-A receptor 3; ETK; ETK1; H TYRO4; HEK; Hek4; Human embryo kinase; human embryo kinase 1; Mek4; Rek4; testicular tissue protein Li 64; TYRO4; TYRO4 protein tyrosine kinase; Tyrosine-protein kinase receptor ETK1; tyrosine-protein kinase receptor REK4; Tyrosine-protein kinase TYRO4
Gene Symbols EPHA3
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human around the phosphorylation site of EPH 3 Tyr779.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13837, 2042, 29210
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.