Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Eph receptor A2/A3 (Tyr588 (A3 Tyr596)) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987827
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7827 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7827 Supplier Bioss Supplier No. BS7554R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Eph receptor A2 is the receptor for members of the ephrin-A family. It binds to ephrin-A1, -A3, -A4 and -A5. Eph receptor A3 is also a receptor for members of the ephrin-A family. It binds to ephrin-A2, -A3, -A4 and -A5 and is thought to play a role in lymphoid function. Eph receptor A4 is a receptor for members of the ephrin-A family. It binds to ephrin-A1, -A4 and -A5. It binds more poorly to ephrin-A2 and -A3. It may play a role in a signal transduction process involved in hindbrain pattern formation.

Specifications

Antigen Phospho-Eph receptor A2/A3 (Tyr588 (A3 Tyr596))
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EPHA2
Gene Accession No. P29317
Gene Symbols EPHA2
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human Eph receptor A2 around the phosphorylation site of Tyr588.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1969
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.