Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Eph receptor B1 (Tyr928) Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS10138R

Description
ephB1, previously known as Elk(eph-like kinase), is a receptor tyrosine kinase of the highly tissue restricted family of eph proteins. EphB1 and other ephB family members are type 1 membrane spanning proteins, comprised of immunoglobulin, fibronectin type III, and cysteine rich subdomains in the ecto domain, and the single uninterrupted cytoplasmic tyrosine kinase domain upstream of a carboxyterminal sterile alpha motif (SAM) domain. ephB family proteins bind ephrins of the B class, ligands that are also transmembrane spanning proteins capable of transmitting signals. ephB1 is expressed predominantly in developing neural structures in embryos, and in vascular epithelium of kidney, and other tissues. Upon binding to alternatively oligomerized ephrin B1, ephB1 signals regulation of cell attachment and cellcell assembly. Members of this protein family are implicated in neuronal and vascular cell targeting.Specifications
Phospho-Eph receptor B1 (Tyr928) | |
Polyclonal | |
Unconjugated | |
EPHB1 | |
9330129L11; AW488255; C130099E04Rik; CAPB; Cek6; DRT; EK6; ELK; Elkh; ELK-related protein tyrosine kinase; ENSMUSG00000074119; Eph receptor B1; Eph receptor B2 (ELK-related protein tyrosine kinase); EPH tyrosine kinase 2; EPHB1; Ephb2; EPHB3; EPH-like kinase 6; Ephrin B1; Ephrin B2; Ephrin B3; Ephrin B4; Ephrin type-B receptor 1; EphrinB1; EphrinB2; EphrinB3; EphrinB4; EPHT2; EPHT3; Epth2; Erk; ETK2; FLJ37986; HEK2; Hek5; HEK6; HEK-6; MGC87492; NET; Neuronally-expressed EPH-related tyrosine kinase; PCBC; soluble EPHB1 variant 1; Tyro5; TYRO6; Tyrosine-protein kinase receptor EPH-2 | |
Rabbit | |
Protein A | |
RUO | |
2047 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P54762 | |
EPHB1 | |
KLH conjugated synthetic phosphopeptide derived from human EphB1 around the phosphorylation site of Tyr928. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction