Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-EphA2/EphA3/EphA4 (Tyr588, Tyr596) Polyclonal Antibody, Invitrogen™

Catalog No. PIPA599134
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599134 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599134 Supplier Thermo Scientific Supplier No. PA599134

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of EPHA2/3/4 only when phosphorylated at Tyrosine 588 and 596.

The Eph subfamily represents the largest group of receptor protein tyrosine kinases identified to date. While the biological activities of these receptors have yet to be determined, there is increasing evidence that they are involved in central nervous system function and in development. The Eph subfamily receptors of human origin (and their murine/avian homologs) include EphA1(Eph), EphA2 (Eck), EphA3 (Hek4), EphA4 (Hek8), EphA5 (Hek7), EphA6 (Hek12),EphA7 (Hek11/MDK1), EphA8 (Hek3), EphB1 (Hek6), EphB2 (Hek5), EphB3(Cek10, Hek2), EphB4 (Htk), EphB5 (Hek9) and EphB6 (Mep). Ligands for Eph receptors include ephrin-A4 (LERK-4) which binds EphA3 and EphB1. In addition, Ephrin-A2 (Elf-1) has been described as the ligand for EphA4, ephrin-A3 (Ehk1-L) as the ligand for EphA5 and ephrin-B2 (Htk-L) as the ligand for EphB4 (Htk).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-EphA2/EphA3/EphA4 (Tyr588, Tyr596)
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EPHA2
Gene Accession No. O08680, P29317, P29319, P29320, P54764, Q03137, Q03145
Gene Alias 2900005C20Rik; AI385584; ARCC2; AW492086; AW545284; Cek4; Cek8; CTPA; CTPP1; CTRCT6; ECK; EK4; EK8; End3; EPH receptor A2; Eph receptor A3; EPH receptor A4; EPHA2; EPHA3; EPHA4; EPH-like kinase 4; EPH-like kinase 8; eph-like tyrosine kinase 1; Ephrin A2; Ephrin A3; Ephrin A4; ephrin receptor EphA2; ephrin type-A receptor 2; Ephrin type-A receptor 3; ephrin type-A receptor 4; EphrinA2; EphrinA3; EphrinA4; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; ETK; ETK1; HEK; HEK4; Hek8; Human embryo kinase; human embryo kinase 1; Mek4; Myk2; rb; receptor protein-tyrosine kinase HEK8; rEK4; RGD1560587; SEK; Sek1; Sek2; Sek-2; soluble EPHA2 variant 1; testicular tissue protein Li 64; Tyro1; TYRO1 protein tyrosine kinase; Tyro4; TYRO4 protein tyrosine kinase; tyrosine-protein kinase receptor ECK; Tyrosine-protein kinase receptor ETK1; tyrosine-protein kinase receptor MPK-3; Tyrosine-protein kinase receptor MPK-5; tyrosine-protein kinase receptor REK4; tyrosine-protein kinase receptor SEK; Tyrosine-protein kinase receptor SEK-1; tyrosine-protein kinase receptor SEK-2; Tyrosine-protein kinase TYRO1; tyrosine-protein kinase TYRO4
Gene Symbols EPHA2, EPHA3, EPHA4
Host Species Rabbit
Immunogen A synthesized peptide derived from human EPHA2(Accession P29317), corresponding to amino acid residues around phosphorylated Tyr588.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13836, 13837, 13838, 1969, 2042, 2043, 29210, 316539, 366492
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.