Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-EphA2/EphA5 (Tyr594) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537812
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537812 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537812 Supplier Invitrogen™ Supplier No. PA537812
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for Western blot is JK cells.

These proteins belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. This gene encodes a protein that binds ephrin-A ligands.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-EphA2/EphA5 (Tyr594)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EPHA5
Gene Accession No. P29317, P54756, Q03145, Q60629
Gene Alias AI854630; ARCC2; AW125296; AW545284; brain-specific kinase; BSK; Cek7; CEK-7; CTPA; CTPP1; CTRCT6; Eck; eck-like sequence 1; Ehk1; EHK-1; EK7; Els1; EPH homology kinase 1; EPH receptor A2; EPH receptor A5; Epha2; EPHA5; EPH-like kinase 7; Ephrin A2; Ephrin A5; Ephrin type-A receptor 2; ephrin type-A receptor 5; EphrinA2; EphrinA5; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; Hek7; Myk2; receptor protein-tyrosine kinase HEK7; Rek7; Sek2; Sek-2; soluble EPHA2 variant 1; TYRO4; Tyrosine-protein kinase receptor ECK; tyrosine-protein kinase receptor EHK-1; tyrosine-protein kinase receptor MPK-5; Tyrosine-protein kinase receptor SEK-2
Gene Symbols EPHA2, EPHA5
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of tyrosine 594 (H-T-Y(p)-E-D) derived from Human EPHA2/5.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13836, 13839, 1969, 2044
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.