Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-EphA3/EphA4/EphA5 (Tyr779, Tyr833) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537813
Description
A suggested positive control for IHC is human brain tissue.
These proteins belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. This gene encodes a protein that binds ephrin-A ligands.
Specifications
Phospho-EphA3/EphA4/EphA5 (Tyr779, Tyr833) | |
Polyclonal | |
Unconjugated | |
EPHA4 | |
2900005C20Rik; AI385584; AI854630; AW125296; AW492086; Brain-specific kinase; bsk; Cek4; CEK7; CEK-7; Cek8; eck-like sequence 1; EHK1; EHK-1; EK4; EK7; EK8; Ekh1; Els1; Els1.; End3; EPH homology kinase 1; Eph receptor A3; Eph receptor A4; EPH receptor A5; EPHA3; EPHA4; Epha5; EPH-like kinase 4; EPH-like kinase 7; EPH-like kinase 8; Eph-like tyrosine kinase 1; Ephrin type-A receptor 3; ephrin type-A receptor 4; Ephrin type-A receptor 5; ETK; ETK1; H TYRO4; HEK; Hek4; Hek7; HEK8; Human embryo kinase; human embryo kinase 1; Mek4; rb; receptor protein-tyrosine kinase HEK7; receptor protein-tyrosine kinase HEK8; rEK4; Rek7; RGD1560587; Sek; Sek1; testicular tissue protein Li 64; TYRO1; TYRO1 protein tyrosine kinase; TYRO4; TYRO4 protein tyrosine kinase; tyrosine-protein kinase receptor EHK-1; tyrosine-protein kinase receptor ETK1; Tyrosine-protein kinase receptor MPK-3; tyrosine-protein kinase receptor REK4; tyrosine-protein kinase receptor SEK; Tyrosine-protein kinase receptor SEK-1; tyrosine-protein kinase TYRO1; Tyrosine-protein kinase TYRO4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2042, 2043, 2044 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P29320, P54756, P54764 | |
EPHA3, EPHA4, Epha5 | |
Peptide sequence around phosphorylation site of tyrosine 779/833 (E-A-Y(p)-T-T)/(Q-D-V(p)-I-K) derived from Human EPHA3/4/5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction