Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-EphA3 (Tyr779) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105554
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105554 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105554 Supplier Invitrogen™ Supplier No. PA5105554
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of EPHA3 only when phosphorylated at Tyr779.

EPHA3 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. EPHA3 encodes a protein that binds ephrin-A ligands and receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EPHA3 binds ephrin-A ligands.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-EphA3 (Tyr779)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EPHA3
Gene Accession No. O08680, P29319, P29320
Gene Alias AW492086; Cek4; EK4; End3; EPH receptor A3; EPHA3; EPH-like kinase 4; Eph-like tyrosine kinase 1; Ephrin type-A receptor 3; ETK; ETK1; H TYRO4; Hek; Hek4; Human embryo kinase; human embryo kinase 1; I79_002835; Mek4; rEK4; testicular tissue protein Li 64; TYRO4; TYRO4 protein tyrosine kinase; tyrosine-protein kinase receptor ETK1; tyrosine-protein kinase receptor REK4; tyrosine-protein kinase TYRO4
Gene Symbols EPHA3
Host Species Rabbit
Immunogen A synthesized peptide derived from human EPHA3(Accession P29320), corresponding to amino acid residues around phosphorylated Tyr779.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13837, 2042, 29210
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.