Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-EphA4 (Tyr779) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143695
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143695 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143695 Supplier Thermo Scientific Supplier No. PA5143695
Add to Cart
Add to Cart

Phospho-EphA4 (Tyr779) Polyclonal Antibody

Phospho-EphA4 (Tyr779) Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-EphA4 (Tyr779)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene EPHA4
Gene Accession No. P54764, Q03137
Gene Alias 2900005C20Rik; AI385584; Cek8; Cek8 protein; EK8; epa4; EPH receptor A4; Epha4; EPH-like kinase 8; Ephrin type-A receptor 4; Hek8; rb; receptor protein-tyrosine kinase HEK8; RGD1560587; Sek; Sek1; Tyro1; TYRO1 protein tyrosine kinase; tyrosine-protein kinase receptor MPK-3; Tyrosine-protein kinase receptor SEK; tyrosine-protein kinase receptor SEK-1; Tyrosine-protein kinase TYRO1
Gene Symbols EPHA4
Host Species Rabbit
Immunogen EphA4 (Tyr-779) synthetic peptide corresponds to amino acids surrounding phosphorylated tyrosine 779 in human EphA4. This sequence has significant homology to the conserved site in rat and mouse EphA4, and is conserved well in most other EphA family members, but has less conservation in EphB family members.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13838, 2043, 316539
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.