Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-EphA7 (Tyr608) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564788
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564788 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564788 Supplier Invitrogen™ Supplier No. PA564788
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-EphA7 (Tyr608) Polyclonal Antibody detects endogenous levels of EphA7 only when phosphorylated at Tyr608.

EPHA7 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. EPHA7 is involved in embryonic development. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-EphA7 (Tyr608)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.87% NaCl, 30% glycerol and 0.01% sodium azide; pH 7.3
Gene EPHA7
Gene Accession No. P54759, Q15375, Q61772
Gene Alias Cek11; Developmental kinase 1; Ebk; EHK3; EHK-3; EK11; embryonic brain kinase; epa7; EPH homology kinase 3; Eph homology kinase-3; EPH receptor A7; Epha7; EPHA7-S; EPH-like kinase 11; ephrin receptor EphA7; ephrin type-A receptor 7; HEK11; Mdk1; mDK-1; receptor protein-tyrosine kinase HEK11; tyrosine-protein kinase receptor EHK-3
Gene Symbols EPHA7
Host Species Rabbit
Immunogen A synthesized peptide derived from human EPHA7 (Phospho-Tyr608).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13841, 171287, 2045
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.