Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-EphA8 (Tyr838) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564576
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564576 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564576 Supplier Invitrogen™ Supplier No. PA564576
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-EphA8 (Tyr838) Polyclonal Antibody detects endogenous levels of EphA8 only when phosphorylated at Tyr838.

EPHA8 belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. EPHA8 plays a role in short-range contact-mediated axonal guidance during development of the mammalian nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-EphA8 (Tyr838)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.87% NaCl, 30% glycerol and 0.01% sodium azide; pH 7.3
Gene Epha8
Gene Accession No. O09127, P29322
Gene Alias AW047546; Eek; EK3; epa8; EPH- and ELK-related kinase; EPH- and ELK-related tyrosine kinase; Eph receptor A8; Epha8; EPH-like kinase 3; Ephrin type-A receptor 8; Ephrin type-A receptor 8 precursor (Tyrosine-protein kinase receptor EEK) (EPH-and ELK-related kinase); Hek3; hydroxyaryl-protein kinase; KIAA1459; mKIAA1459; protein-tyrosine kinase; tyrosine-protein kinase receptor EEK; tyrosylprotein kinase
Gene Symbols Epha8
Host Species Rabbit
Immunogen A synthesized peptide derived from human EPHA8 (Phospho-Tyr838).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13842, 2046
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.