Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-EphB1/EphB2 (Tyr778, Tyr780) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106133
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106133 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106133 Supplier Invitrogen™ Supplier No. PA5106133
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of EPHB1/2 only when phosphorylated at Tyr778/Tyr780.

Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase family. The protein encoded by this gene is a receptor for ephrin-B family members.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-EphB1/EphB2 (Tyr778, Tyr780)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EPHB1
Gene Accession No. P09759, P29323, P54762, P54763, Q8CBF3
Gene Alias 9330129L11; AW488255; C130099E04Rik; CAPB; Cek5; Cek6; Developmentally-regulated Eph-related tyrosine kinase; Drt; EK5; EK6; ELK; Elkh; ELK-related protein tyrosine kinase; elk-related tyrosine kinase; ENSMUSG00000074119; Eph receptor B1; Eph receptor B2; Eph receptor B2 (ELK-related protein tyrosine kinase); eph tyrosine kinase 2; EPH tyrosine kinase 3; Ephb1; EPHB2; EphB2/CTF1; EphB2/CTF2; EPHB3; EPH-like kinase 5; EPH-like kinase 6; Ephrin B1; Ephrin B2; Ephrin type-B receptor 1; ephrin type-B receptor 2; EphrinB1; EphrinB2; EPHT2; EPHT3; Epth2; Epth3; ERK; ETECK; ETK2; FLJ37986; HEK2; Hek5; Hek6; HEK-6; MGC87492; NET; neural kinase; Neuronally-expressed EPH-related tyrosine kinase; Nuk; Nuk receptor tyrosine kinase; PCBC; Prkm5; protein-tyrosine kinase HEK5; Qek5; Renal carcinoma antigen NY-REN-47; RGD1564232; Sek3; soluble EPHB1 variant 1; Tyro5; TYRO6; Tyrosine-protein kinase receptor EPH-2; tyrosine-protein kinase receptor EPH-3; Tyrosine-protein kinase receptor SEK-3; tyrosine-protein kinase TYRO5
Gene Symbols EPHB1, EPHB2
Host Species Rabbit
Immunogen A synthesized peptide derived from human EPHB1(Accession P54762), corresponding to amino acid residues around phosphorylated Tyr778.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13844, 2047, 2048, 24338, 270190, 313633
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.