Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Ephrin B1 (Tyr313, Tyr317) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105608
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105608 Supplier Invitrogen™ Supplier No. PA5105608
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of EFNB1 only when phosphorylated at Tyr313/317.

Cell surface transmembrane ligand for Eph receptors, a family of receptor tyrosine kinases which are crucial for migration, repulsion and adhesion during neuronal, vascular and epithelial development. Binds promiscuously Eph receptors residing on adjacent cells, leading to contact-dependent bidirectional signaling into neighboring cells. The signaling pathway downstream of the receptor is referred to as forward signaling while the signaling pathway downstream of the ephrin ligand is referred to as reverse signaling. Binds to receptor tyrosine kinase including EPHA4, EPHA3 and EPHB4. Together with EPHB4 plays a central role in heart morphogenesis and angiogenesis through regulation of cell adhesion and cell migration. EPHB4-mediated forward signaling controls cellular repulsion and segregation from EFNB2-expressing cells. May play a role in constraining the orientation of longitudinally projecting axons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Ephrin B1 (Tyr313, Tyr317)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EFNB1
Gene Accession No. P52795, P52796, P98172
Gene Alias CEK5 ligand; CEK5 receptor ligand; CEK5-L; CEL5-L; CFND; CFNS; EFB1; EFL3; EFL-3; EFL6; EFNB1; EFNB3; ELK ligand; ELK-L; EPH-related receptor tyrosine kinase ligand 2; ephrin B1; Ephrin B3; EphrinB1; ephrin-B1; Ephrin-B1 C-terminal fragment; Ephrin-B1 CTF; Ephrin-B1 ICD; Ephrin-B1 intracellular domain; Ephrin-B1 precursor; Epl2; EPLG2; EPLG8; kinase ephrin-B1; Lerk2; LERK-2; LERK8; Stimulated by retinoic acid gene 1 protein; Stra1; STRA1 protein; transmembrane ligand
Gene Symbols EFNB1
Host Species Rabbit
Immunogen A synthesized peptide derived from human EFNB1(Accession P98172), corresponding to amino acid residues around phosphorylated Tyr313.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13641, 1947, 25186
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.