Learn More
Invitrogen™ Phospho-Ephrin B2 (Tyr330) Polyclonal Antibody

Description
Antibody detects endogenous levels of Ephrin B2 only when phosphorylated at Tyrosine 330.
Specifications
Specifications
| Antigen | Phospho-Ephrin B2 (Tyr330) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Efnb2 |
| Gene Accession No. | P52799, P52800 |
| Gene Alias | EFNB2; efnb2a; Elf2; ELF-2; EPH-related receptor tyrosine kinase ligand 5; ephrin B2; ephrin B2a; ephrin-B2; Ephrin-B2a; Epl5; EPLG5; HTK ligand; Htkl; HTK-L; id:ibd5072; Lerk5; LERK-5; ligand of eph-related kinase 5; MGC126226; MGC126227; MGC126228; NLERK-1 |
| Gene Symbols | Efnb2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.