Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-EPO Receptor (Tyr485) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982602
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2602 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2602 Supplier Bioss Supplier No. BS13088R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recombinant Human Erythropoietin Receptor (EPO R) is a type I membrane-spanning protein, expressed in a mouse myeloma cell line, NSO. This product is produced from a cDNA sequence encoding the extracellular domain of the EPO receptor. The 225 amino acid residue mature recombinant soluble EPO receptor, generated after cleavage of a 25 amino acid residue signal peptide, has predicted molecular mass of approximately 25 kDa.

Specifications

Antigen Phospho-EPO Receptor (Tyr485)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EPOR
Gene Accession No. P19235
Gene Alias EPO R; EPOR; EPO-R; Erythropoietin receptor; pEpoR
Gene Symbols EPOR
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human EPO Receptor around the phosphorylation site of Tyr485.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2057
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.